Learn More
Hach Company Manganese Reagent Set, HR, 25 mL, Periodate Oxidation
Supplier:  Hach Company 2243200
Encompass
For determination of Manganese by the Periodate Oxidation Method, Hach Method 8034. EPA Approved for wastewater reporting (digestion required) - Federal Register, 44, (116) 34193 (June 14, 1979). Range: 0.1 - 20.0 mg/l Mn. Sample Size: 25 mL. Set Contains: Buffer Powder Pillows (Citrate type) and Sodium Periodate Powder Pillows for 100 tests.
Catalog No. 50-199-5618