Learn More
Hach Company Molybdenum Reagent Set, HR, 10 mL
Supplier:  Hach Company 2604100
Encompass
For determination of Molybdate Molybdenum by the Mercaptoacetic Acid Method. Hach method 8036. Sample Size: 10 mL. Range: 0.3 - 40.0 mg/L Mo. Reagent set includes MolyVer 1, 2 and 3 Molybdenum Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-6321