Learn More
Hach Company Molybdenum Reagent Set, LR, 50 mL
Supplier:  Hach Company 2440200
Encompass
For determination of Low Range Molybdate Molybdenum by the Ternary Complex method (Hach method 8169). Range: 0.02 - 3.00 mg/L as Mo. Sample Size: 50 mL. Set contains: Molybdenum 1 Reagent Powder Pillows and Molybdenum 2 Reagent Solution for approximately 100 tests.
Catalog No. 50-199-6003