Learn More
Hach Company LR Molybdenum Reagent Set, 5 mL
Supplier:  Hach Company 2439500
Encompass
For determination of Low Range Molybdate Molybdenum by the ternary complex method, using visual test kits. Replacement reagent set for MO-LR Test Kit (2359300). Range: 0.25 - 3 mg/L Mo. Sample Size: 5 mL. Set contains: Molybdenum 1 Reagent Powder Pillows and Molybdenum 2 Reagent Solution for approximately 100 tests.
Catalog No. 50-199-5999