Learn More
Hach Company Molybdenum Reagent Set, LR, 20mL
Supplier:  Hach Company 2449400
Encompass
For determination of low range Molybdate Molybdenum by the Ternary Complex method. Hach Method 8169. Range: 0.02 - 3.00 mg/L Mo Sample Size: 20 mL Reagent Set includes: Molybdenum 1 Reagent (LR) Molybdate Powder Pillows and Molybdenum 2 Reagent Solution for approximately 100 tests.
Catalog No. 50-199-6032