Learn More
Hach Company Nitrogen-Nitrate Reagent Set, LR, 10 mL, Cadmium Reduction
Catalog No. 501995971 Shop All Hach Company Products
Encompass
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach Method 8192. Range: 0.01 - 0.50 mg/L NO3-N. Sample Size: 10 mL. Set contains NitraVer 6 and NitriVer 3 Nitrite Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5971 Supplier Hach Company Supplier No. 2429800
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach Method 8192. Range: 0.01 - 0.50 mg/L NO3-N. Sample Size: 10 mL. Set contains NitraVer 6 and NitriVer 3 Nitrite Reagent Powder Pillows for 100 tests.