Learn More
Hach Company Nitrogen-Nitrate Reagent Set, LR, 5 mL Sample
Supplier:  Hach Company 2460800
Encompass
For determination of Nitrogen-Nitrate by the Cadmium Reduction method. Sample Size: 5 mL For visual test kits. Reagent set includes: NitriVer 3 Powder Pillows and NitraVer 6 Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-6040