Learn More
Hach Company Nitrogen-Nitrate Reagent Set, LR, 25 mL, Cadmium Reduction
Catalog No. 501995634 Shop All Hach Company Products
Encompass
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach Method 8192. Range: up to 0.50 mg/L NO3-N. Sample Size: 25 mL. Reagent Set includes: NitriVer 3 Reagent Powder Pillows and NitraVer 6 Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5634 Supplier Hach Company Supplier No. 2245100
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach Method 8192. Range: up to 0.50 mg/L NO3-N. Sample Size: 25 mL. Reagent Set includes: NitriVer 3 Reagent Powder Pillows and NitraVer 6 Reagent Powder Pillows for 100 tests.