Learn More
Hach Company Phosphonate Reagent Set, 10 mL
Supplier:  Hach Company 2429700
Encompass
For determination of Phosphonate compounds by the UV Persulfate Oxidation Method. Hach Method 8007. Ranges: 0.02 - 2.50 to 1 - 125 mg/L. Sample Size: 10 mL Set includes PhosVer™ 3 Phosphate Reagent Powder Pillows and Potassium Persulfate Powder Pillows for 100 tests.
Catalog No. 50-199-5970