Learn More
Hach Company Phosphonate Reagent Set, 25 mL
Supplier: Hach Company 2244000
For determination of Phosphonates by the Persulfate/UV Oxidation Method. Hach method 8007. Ranges: 0.02 - 2.50 mg/L to 1 - 125 mg/L. Sample Size: 25 mL. Reagent Set includes: PhosVer 3 Phosphate Reagent Powder Pillows and Potassium Persulfate Powder Pillows for 100 tests.