Learn More
Hach Company Phosphorus, Reactive, Reagent Set, Amino Acid
Catalog No. 501995626 Shop All Hach Company Products
Encompass
For determination of Reactive (Ortho-) Phosphorus (Phosphate) by the Amino Acid Method. Hach Method 8178. Range: 0.23 - 30.00 mg/L PO4. Sample Size: 25 mL Set includes: Amino Acid Reagent and Molybdate Reagent for 100 tests.
Exact match for part number 2244100
Catalog No. 50-199-5626 Supplier Hach Company Supplier No. 2244100
For determination of Reactive (Ortho-) Phosphorus (Phosphate) by the Amino Acid Method. Hach Method 8178. Range: 0.23 - 30.00 mg/L PO4. Sample Size: 25 mL Set includes: Amino Acid Reagent and Molybdate Reagent for 100 tests.
Exact match for part number 2244100