Learn More
Hach Company Phosphorus Digestion Reagent Set
Supplier:  Hach Company 2459000
Encompass
For acid persulfate digestion of samples for Total Phosphorus (Phosphate) determination by Hach photometric methods. Requires additional reagents for measurement of the reactive phosphorus generated by the digestion. Set contains Potassium Persulfate Powder Pillows, Sodium Hydroxide Solution (5.0 N), and Sulfuric Acid (5.25 N). Approximately 100 tests.
Catalog No. 50-199-6034