Learn More
Hach Company Quaternary Ammonium Compound Reagent Set
Supplier:  Hach Company 2459200
Encompass
Reagent set for determination of Quaternary Ammonium Compounds by the Direct Binary Complex Method. Hach Method 8337. Range: 0.2 - 5.0 mg/L as Cetyltrimethyl Ammonium Bromide (CTAB). This test is applicable to the monitoring of QAC's in swimming pools and cooling towers. Reagent set contains QAC Reagent 1 and QAC Reagent 2 Powder Pillows for 100 tests.
Catalog No. 50-199-6036