Learn More
Hach Company Silica Reagent Set, HR, 5mL Samples, 100 Tests
Supplier:  Hach Company 2462300
Encompass
For high range silica determination by the Silicomolybdate method. For visual test kits that require a 5 mL sample. 100 tests
Catalog No. 50-199-6041