Learn More
Hach Company Silica Reagent Set, High Range, 10 mL
Supplier:  Hach Company 2429600
Encompass
For High Range Silica determination by the Silicomolybdate method. Hach Method 8185. Range: 1 - 100 mg/L SiO2. Sample Size: 10 mL. Set contains Acid Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5969