Learn More
Hach Company Silica Reagent Set, LR Silica, 25 mL
Supplier:  Hach Company 2346800
Encompass
For determination of low range silica by the heteropoly blue method. Hach method 8186 Sample Size: 25 mL, Range: 0.01 to 1.60 mg/L, 100 Tests
Catalog No. 50-199-5838