Learn More
Hach Company Silica Reagent Set, Ultra Low Range, 40 tests
Supplier:  Hach Company 2581400
Encompass
For ultralow range silica determination by the Heteropoly Blue method. Ampuled Amino Acid F Reagent for stability.
Catalog No. 50-199-6262