Learn More
Hach Company Sulfite Reagent Set for SU-5 Kit
Supplier:  Hach Company 2440000
Encompass
Reagent set for determination of Sulfite by iodometric drop count titration. Replacement reagent set for SU-5 Test Kit (148002). Range: Up to 200 mg/L SO3. Set includes Sulfamic Acid Powder Pillows, Sulfite 1 Reagent Powder Pillows and Sulfite 3 Solution for approximately 100 tests.
Catalog No. 50-199-6002