Learn More
Hach Company Sulfite Reagent Set, Buret Titration
Supplier:  Hach Company 2459800
Encompass
For determination of Sulfite by iodometric buret titration. Hach Method 8071 Range: 0 to 500+ mg/L SO3. Reagent set includes: Dissolved Oxygen 3 Reagent Powder Pillows, Potassium Iodide-Iodate Standard Solution and Starch Indicator Solution for approximately 100 tests.
Catalog No. 50-199-6039