Learn More
Hach Company Sulfite Reagent Set, Digital Titrator
Catalog No. 501995694 Shop All Hach Company Products
Encompass
For determination of sulfite by iodate-iodide Digital Titrator titration, Hach Method 8216. Range: 4 to 400+ mg/L SO3. Reagent Set includes: Dissolved Oxygen 3 Reagent Powder Pillows, Iodate-Iodide Cartridge ( 0.3998 N), and Starch Indicator. Approximately 40-100 tests
Catalog No. 50-199-5694 Supplier Hach Company Supplier No. 2272300
For determination of sulfite by iodate-iodide Digital Titrator titration, Hach Method 8216. Range: 4 to 400+ mg/L SO3. Reagent Set includes: Dissolved Oxygen 3 Reagent Powder Pillows, Iodate-Iodide Cartridge ( 0.3998 N), and Starch Indicator. Approximately 40-100 tests