Learn More
Hach Company ToxTrak™ Toxicity Reagent Set, 25-49 tests
Supplier:  Hach Company 2597200
Encompass
Use the ToxTrak™ Toxicity Reagent Set to analyze your water or wastewater samples for toxicity. Follows Hach Toxicity Method 10017.
Catalog No. 50-199-6297