Learn More
Hach Company Chlorine Reagent Set (Total), Ultra Low Range DPD
Supplier:  Hach Company 2563000
Encompass
For determination of ultralow Total Chlorine concentrations by the DPD Method. Hach Method 8370. USEPA accepted for drinking water and wastewater analysis. Range: 2 - 500 μg/L Cl₂. Reagents are ampuled for stability. Reagent set includes ULR Chlorine Buffer Solution (1.5 mL ampules), DPD Indicator Solution for ULR Chlorine (1.5 mL ampules) and Blanking Reagent for 20 tests.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6234