Learn More
Alkali Scientific ASI™ Reagent Solution Reservoir, 7mL, 100/cs
SDP
Catalog No. 503026907
Encompass_Preferred
Single and multi-channel reservoirs for cell culture work. Wells have notched volume labels and sloping sides for easy pipetting. Volumes refer to each individual channel. 7 mL, 8 channel, individually wrapped, sterile, 100 per case
Catalog No. 50-302-6907 Supplier Alkali Scientific Supplier No. RS671
Single and multi-channel reservoirs for cell culture work. Wells have notched volume labels and sloping sides for easy pipetting. Volumes refer to each individual channel. 7 mL, 8 channel, individually wrapped, sterile, 100 per case