Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Life Sciences Reagent Stock Solution

Catalog No. 06713245
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-713-245 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-713-245 Supplier Beckman Coulter Life Sciences Supplier No. 9459400
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The reagent stock solution is a concentrated mixture of acid and persulfate; a working solution is simply a 1:100 dilution of the stock solution.

The reagent stock solution is a concentrated mixture of acid and persulfate; a working solution is simply a 1:100 dilution of the stock solution.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonim Persulfate, Phosphoric Acid
CAS 7727-54-0, 7664-38-2
Percent Purity 20 to 30%, <10%
Packaging Glass Bottle
pH 0.5
Quantity 500 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.