Learn More
McKesson General Medical REAGENT STROMATOLYSER,500ML,3/
Catalog No. NC2128841
Encompass_Preferred
Reagent Stromatolyser®-WH Hematology HGB / WBC Count For Sysmex Automated Hematology Analyzers 3 X 500 mLREAGENT, STROMAT 500ML (3/BX)
Catalog No. NC2128841 Supplier McKesson General Medical Supplier No. 1204755
Reagent Stromatolyser®-WH Hematology HGB / WBC Count For Sysmex Automated Hematology Analyzers 3 X 500 mLREAGENT, STROMAT 500ML (3/BX)