Learn More
Hach Company Hach Reagent Delivery Valve for 5500sc Analyzer (6783700)
Catalog No. NC2590878 Shop All Hach Company Products
Encompass
This replacement reagent delivery valve (MPN 6783700) is a service part for Hach 5500sc analyzers used to control reagent dispensing in automated analysis. Install as a replacement when leaks or incorrect dispense volumes are detected.
  • Controls reagent flow during analysis.
  • Used as a replaceable maintenance part in the analyzer reagent delivery system.
  • Replace if valve leaks or dispenses incorrect volume.
  • Diagnostic test sets each valve to supply 2,000 μL (2 mL) to verify performance.
  • Sold as a single replacement unit; dimensions not specified in manufacturer documentation.
Catalog No. NC2590878 Supplier Hach Company Supplier No. 6783700
This replacement reagent delivery valve (MPN 6783700) is a service part for Hach 5500sc analyzers used to control reagent dispensing in automated analysis. Install as a replacement when leaks or incorrect dispense volumes are detected.
  • Controls reagent flow during analysis.
  • Used as a replaceable maintenance part in the analyzer reagent delivery system.
  • Replace if valve leaks or dispenses incorrect volume.
  • Diagnostic test sets each valve to supply 2,000 μL (2 mL) to verify performance.
  • Sold as a single replacement unit; dimensions not specified in manufacturer documentation.