Learn More
Leica Biosystems Reagent Vessel With Handle, 1.8 L for Leica TP1020 Tissue Processor
Catalog No. NC3359554
Encompass
Leica Biosystems reagent vessel with handle (MPN 14 0422 42545) is a 1.8 L reagent container intended for use in automated tissue processors such as the TP1020. It provides a secure, removable vessel for holding processing reagents during tissue processing.
  • Holds 1.8 L of reagent.
  • Includes an integrated handle for safe lifting and removal.
  • Designed for use with automated tissue processors to contain processing reagents.
Catalog No. NC3359554 Supplier Leica Biosystems Supplier No. 14042242545
Leica Biosystems reagent vessel with handle (MPN 14 0422 42545) is a 1.8 L reagent container intended for use in automated tissue processors such as the TP1020. It provides a secure, removable vessel for holding processing reagents during tissue processing.
  • Holds 1.8 L of reagent.
  • Includes an integrated handle for safe lifting and removal.
  • Designed for use with automated tissue processors to contain processing reagents.