Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents for Electrophoresis: UltraSpec-Agarose™
SDPSCI_ED

Catalog No. S11605B Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
20 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11605B 100 g
S11605 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11605B Supplier Edvotek™ Supplier No. 605100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Edvotek™ Reagents for Electrophoresis: UltraSpec-Agarose™ DNA Electrophoresis Grade reagent.

DNA Electrophoresis Grade

Specifications

Color White
Format Melt and Pour
Separation of DNA
Separation Type Electrophoresis
For Use With (Application) Agarose or Polyacrylamide Gel electrophoresis
Material Agarose
Quantity 100 g
Product Type Agarose Gel
For Use With (Equipment) Mini Gel Tank
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.