Learn More
Medchemexpress LLC Real Thiol | 2280796-90-7 | 98.37% | 411.36 | 1 MG
SDP
Catalog No. 5000365588
Encompass_Preferred
Real Thiol is a reversible reaction-based fluorescent probe which can quantitatively monitor the real-time glutathione dynamics in living cells.
  • Provides ratiometric fluorescence responses with a wide dynamic range when reacting with glutathione.
  • Has improved quantum yields and photostability.
  • Allows monitoring of glutathione level changes within 1 to 10 mM in both directions in living cells.
Catalog No. 50-003-65588 Supplier Medchemexpress LLC Supplier No. 1MGHY108715
Real Thiol is a reversible reaction-based fluorescent probe which can quantitatively monitor the real-time glutathione dynamics in living cells.
  • Provides ratiometric fluorescence responses with a wide dynamic range when reacting with glutathione.
  • Has improved quantum yields and photostability.
  • Allows monitoring of glutathione level changes within 1 to 10 mM in both directions in living cells.