Learn More
Andwin Scientific REAL-TIME PCR STRIP TUBES
SDP
Catalog No. NC0104766
Encompass_Preferred
The 0,2 mL GeneMate SnapStrip 8-Strip Standard PCR Tubes feature individually attached flat caps that helps in the transmission and also acts as a writing or marking surface. The caps also have a high clarity manufacturing that is ideal for real-time PCR (QPCR) applications where excitation and emission takes through the cap. The tubes feature great compatibility with a wide range of thermal cyclers.
Catalog No. NC0104766 Supplier Andwin Scientific Supplier No. T31961
The 0,2 mL GeneMate SnapStrip 8-Strip Standard PCR Tubes feature individually attached flat caps that helps in the transmission and also acts as a writing or marking surface. The caps also have a high clarity manufacturing that is ideal for real-time PCR (QPCR) applications where excitation and emission takes through the cap. The tubes feature great compatibility with a wide range of thermal cyclers.