Learn More
Alkali Scientific ASI™ Optical qPCR Sealing Film, 100/cs
SDP
Catalog No. 502336036
Encompass_Preferred
With the highest transparency, this polyolefin film offers a special pressure sensitive DMSO-resistant silicone adhesive embedded inside the film itself. A polyester backing with end tabs assures easy positioning of the plate. This film is also perfectly suited for raised-rim plates ensuring reliable sealing around each well. Functional from -40 °C to +120 °C. Certified RNAse, DNAse and DNA-free. 100 per pack.
Catalog No. 50-233-6036 Supplier Alkali Scientific Supplier No. SF6ML
With the highest transparency, this polyolefin film offers a special pressure sensitive DMSO-resistant silicone adhesive embedded inside the film itself. A polyester backing with end tabs assures easy positioning of the plate. This film is also perfectly suited for raised-rim plates ensuring reliable sealing around each well. Functional from -40 °C to +120 °C. Certified RNAse, DNAse and DNA-free. 100 per pack.