Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ RealTorqueDigital™ Overhead Stirrers 220V

Catalog No. 14501085 Shop All Fisherbrand Products
Change view
Click to view available options
Speed:
20 to 3000 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Speed
14-501-085 20 to 3000 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 14-501-085
Get a Reusable Glass Beaker with Stirrer Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

High-torque, digital overhead stirrer offering speed and torque capabilities for a wide range of volumes and viscosities

  • Description
  • Stirrer handles high-volume/high-viscosity stirring as easily as homogenizing and dispersing - especially useful for continuous use
  • User-friendly keypad permits precise speed adjustment.
  • Electronic controls permit speed adjustment to single-digit increments and maintain set speed within 1% as solution viscosity changes
  • Rugged, sealed metal housing encloses the high-performance brushless DC motor with automatic overload protection; and two-speed transmission that electronically shifts between high-torque and high-speed range - actually transferring motor's power to chuck with 95% efficiency
  • Chuck is adjustable up to 3/8 in. (10mm)
  • Chuck guard protects chuck from splashes and prevents accidental movement of chuck
  • Stirrer features electronic controls
  • Order Info
  • Dimensions: 8L x 33/4W x 9 in.H (20.3 x 9.5 x 22.9cm). Weight: 11 lb. (5.0kg). Includes chuck, chuck key, chuck guard, support rod and 2 cord sets (UK and EU). Note: does not include impeller, clamp or stand.
  • Certifications
  • Product is CE marked; RoHS and WEEE Compliant.
  • Warranty and Services
  • Three-year

Specifications

Speed 20 to 3000 rpm
Capacity 60 L
Display Digital
Voltage 220/240 V
Diameter (English) Shaft 0.37 in.
Diameter (Metric) Shaft 10 mm
Dimensions (L x W x H) 20.3 x 9.5 x 22.9 cm
Height (English) 9 in.
Height (Metric) 22.9 cm
Length (English) 8 in.
Length (Metric) 20.3 cm
Frequency 50/60 Hz
Horsepower 1/5 hp
Certifications/Compliance CSA, CE, IEC, EMC RoHS, WEE
For Use With (Application) Larger batches of buffers and media to lotions and slurries, this stirrer will exceed your expectations. Ideal for workplaces with changing mixing needs as the business evolves
Includes Overhead stirrer motor, chuck, chuck guard, chuck key, support rod, power cord, manual in four languages (English, French, Spanish and German), calibration certificate and three year warranty
Material Totally Enclosed Metal Housing (TENV)
Motor Type Brushless DC
Torque 341 Ncm
Type Overhead Stirrer
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.