Learn More
MCR Safety Reaper™ 3 -Black Frame, MAX6™ Lens, Closed Cell Foam Gasket; vented eyewear; seals out debris;
Shop All MCR Safety ProductsDescription
- Interchangeable Temples & Adjustable Strap
- Removable Closed Cell Foam Lined Gasket
- MAX6™ Premium Anti-Fog Coating
- Dielectric Frame Design
- Soft TPR Nosepads and Temples Tips
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| For Use With (Application) | Manufacturing |
| Frame Color | Black |
| No. per Pack | 1 Pair |
| Temple Style | Interchangeable Temples & Adjustable Strap |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| UV Protection | UV400 |