Learn More
MCR Safety Reaper™ 3 -Black Frame, MAX6™ Lens, Closed Cell Foam Gasket; vented eyewear; seals out debris;
Catalog No. 19812654 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Tint:
Brown
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-812-654 Brown
19-812-651 Clear
19-812-652 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-812-654 Supplier MCR Safety Supplier No. RP31BPF
This item is not returnable. View return policy
Forms a seal around each eye blocking out debris and small particles. Each side is vented to allow air to flow inside the lens. This product meets the ANSI Z87+ and CSA Z94.3 standards for eyewear.
  • Interchangeable Temples & Adjustable Strap
  • Removable Closed Cell Foam Lined Gasket
  • MAX6™ Premium Anti-Fog Coating
  • Dielectric Frame Design
  • Soft TPR Nosepads and Temples Tips
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog
Lens Tint Brown
For Use With (Application) Manufacturing
Frame Color Black
No. per Pack 1 Pair
Temple Style Interchangeable Temples & Adjustable Strap
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400