Learn More
Medchemexpress LLC Rebamipide-d4 | 1219409-06-9 | 99.0% | 1 MG
SDP
Catalog No. 5000406882
Encompass_Preferred
Rebamipide-d4 is deuterium labeled Rebamipide, a mucoprotective agent. It induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06882 Supplier Medchemexpress LLC Supplier No. HYB0360S1MG
Rebamipide-d4 is deuterium labeled Rebamipide, a mucoprotective agent. It induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.