Learn More
Cayman Chemical Rebamipide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1219409-06-9, 1 mg
SDP
Catalog No. 502950043
Encompass_Preferred
An internal standard for the quantification of rebamipide by GC- or LC-MS
Catalog No. 50-295-0043 Supplier Cayman Chemical Supplier No. 335431 MG
An internal standard for the quantification of rebamipide by GC- or LC-MS