Learn More
Medchemexpress LLC HY-13024 10mg , Rebastinib CAS:1020172-07-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302410MG
Encompass_Preferred
HY-13024 10mg Rebastinib CAS: 1020172-07-9 Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9679