Learn More
Cayman Chemical Rebaudioside A, Purity: ≥98%, CAS: 58543-16-1, 10 g
SDP
Catalog No. 502953136
Encompass_Preferred
A natural non-caloric sweetener; increases GLP-1 secretion in a 2-D mouse intestine model; mice prefer rebaudioside A water to unsweetened water
Catalog No. 50-295-3136 Supplier Cayman Chemical Supplier No. 1189410 G
A natural non-caloric sweetener; increases GLP-1 secretion in a 2-D mouse intestine model; mice prefer rebaudioside A water to unsweetened water