Learn More
Selleck Chemical LLC Rebaudioside A S3796-25mg
SDP
Catalog No. 503110530
Encompass_Preferred
Rebaudioside A (Stevia Extract Reb A Glycoside A3 Stevioside A3) an natural sweetening ingredient extracted from the stevia leaf is used in powder or liquid form to heighten sweetening intensity and improve taste Rebaudioside A is a steviol glycoside -glucosidase inhibitor with IC50 of 35 01 g/ml can inhibit ATP-sensitive K -channels
Catalog No. 50-311-0530 Supplier Selleck Chemical LLC Supplier No. S379625MG
Rebaudioside A (Stevia Extract Reb A Glycoside A3 Stevioside A3) an natural sweetening ingredient extracted from the stevia leaf is used in powder or liquid form to heighten sweetening intensity and improve taste Rebaudioside A is a steviol glycoside -glucosidase inhibitor with IC50 of 35 01 g/ml can inhibit ATP-sensitive K -channels