Learn More
Medchemexpress LLC Rebaudioside A 25g
SDP
Catalog No. 5000669546
Encompass_Preferred
Rebaudioside A is an orally effective steviol glycoside with high sweetness Rebaudioside A acts as an inhibitor of -glucosidase with an IC50 value of 35 01 g/mL Rebaudioside A increases the ATP/ADP ratio in cells in a glucose-dependent manner thereby inhibiting KATP channels leading to membrane depolarization calcium influx and ultimately stimulating insulin secretion Rebaudioside A activates the SREBP signaling pathway by inhibiting HMGCR the rate-limiting enzyme in cholesterol synthesis resulting in increased expression of LDLR on the cell surface thus promoting the uptake of LDL-C in the blood Rebaudioside A can be used for studies on blood glucose and lipid regulation as well as anti-obesity[1][2][3][4][5]
Catalog No. 50-006-69546 Supplier Medchemexpress LLC Supplier No. 25GHYN0466
Add to Cart
Add to Cart
Rebaudioside A is an orally effective steviol glycoside with high sweetness Rebaudioside A acts as an inhibitor of -glucosidase with an IC50 value of 35 01 g/mL Rebaudioside A increases the ATP/ADP ratio in cells in a glucose-dependent manner thereby inhibiting KATP channels leading to membrane depolarization calcium influx and ultimately stimulating insulin secretion Rebaudioside A activates the SREBP signaling pathway by inhibiting HMGCR the rate-limiting enzyme in cholesterol synthesis resulting in increased expression of LDLR on the cell surface thus promoting the uptake of LDL-C in the blood Rebaudioside A can be used for studies on blood glucose and lipid regulation as well as anti-obesity[1][2][3][4][5]