Learn More
Medchemexpress LLC Rebaudioside A | 58543-16-1 | 967.01 | 5 G
SDP
Catalog No. 5000389043
Encompass_Preferred
Rebaudioside A is an orally effective steviol glycoside characterized by its high sweetness. It functions as an α-glucosidase inhibitor and enhances the ATP/ADP ratio in β cells in a glucose-dependent manner, stimulating insulin secretion. It also activates the SREBP signaling pathway by inhibiting HMGCR, increasing LDLR expression and promoting LDL-C uptake. Rebaudioside A is suitable for studies related to blood glucose and lipid regulation, as well as anti-obesity research.
  • Acts as an inhibitor of α-glucosidase
  • Increases ATP/ADP ratio in β cells
  • Stimulates insulin secretion
  • Activates SREBP signaling pathway
  • Promotes uptake of LDL-C
  • Suitable for blood glucose and lipid regulation studies
  • Suitable for anti-obesity research
Catalog No. 50-003-89043 Supplier Medchemexpress LLC Supplier No. HYN04665G
Rebaudioside A is an orally effective steviol glycoside characterized by its high sweetness. It functions as an α-glucosidase inhibitor and enhances the ATP/ADP ratio in β cells in a glucose-dependent manner, stimulating insulin secretion. It also activates the SREBP signaling pathway by inhibiting HMGCR, increasing LDLR expression and promoting LDL-C uptake. Rebaudioside A is suitable for studies related to blood glucose and lipid regulation, as well as anti-obesity research.
  • Acts as an inhibitor of α-glucosidase
  • Increases ATP/ADP ratio in β cells
  • Stimulates insulin secretion
  • Activates SREBP signaling pathway
  • Promotes uptake of LDL-C
  • Suitable for blood glucose and lipid regulation studies
  • Suitable for anti-obesity research