Learn More
Medchemexpress LLC Rebaudioside B | 58543-17-2 | 5 MG
SDP
Catalog No. 5000418244
Encompass_Preferred
Rebaudioside B is a minor constituent isolated from the leaves of Stevia rebaudiana Bertoni. It is approximately 150 times sweeter than sucrose and can undergo hydrolysis to steviol. This product is for research use only.
  • Isolated from Stevia rebaudiana Bertoni
  • Approximately 150 times sweeter than sucrose
  • Can undergo hydrolysis to steviol
Catalog No. 50-004-18244 Supplier Medchemexpress LLC Supplier No. HYN68085MG
Rebaudioside B is a minor constituent isolated from the leaves of Stevia rebaudiana Bertoni. It is approximately 150 times sweeter than sucrose and can undergo hydrolysis to steviol. This product is for research use only.
  • Isolated from Stevia rebaudiana Bertoni
  • Approximately 150 times sweeter than sucrose
  • Can undergo hydrolysis to steviol