Learn More
Cayman Chemical RebaudIosIde C 10mg
SDP
Catalog No. 502954763
Encompass_Preferred
A natural non-caloric sweetener; one of the predominant steviol glycosides from S. rebaudiana; metabolized by gut microbiota to steviol
Catalog No. 50-295-4763 Supplier Cayman Chemical Supplier No. 1189510 MG
A natural non-caloric sweetener; one of the predominant steviol glycosides from S. rebaudiana; metabolized by gut microbiota to steviol