Learn More
Selleck Chemical LLC Rebaudioside C S4742-2mg
SDP
Catalog No. 503108429
Encompass_Preferred
Rebaudioside C (Dulcoside B) belongs to the family of Steviol Glycosides It is a natural constituent of the plant Stevia rebaudiana Bertoni and used as natural sweeteners to diabetics and others on carbohydrate-controlled diets
Catalog No. 50-310-8429 Supplier Selleck Chemical LLC Supplier No. S47422MG
Rebaudioside C (Dulcoside B) belongs to the family of Steviol Glycosides It is a natural constituent of the plant Stevia rebaudiana Bertoni and used as natural sweeteners to diabetics and others on carbohydrate-controlled diets