Learn More
Cayman Chemical Rebaudioside D, Purity: ≥95%, CAS: 63279-13-0, 10 mg
SDP
Catalog No. 502937129
Encompass_Preferred
A natural non-caloric sweetener; a steviol glycoside that has been isolated from S. rebaudiana leaves; metabolized to steviol by gut microbiota
Catalog No. 50-293-7129 Supplier Cayman Chemical Supplier No. 2788010 MG
A natural non-caloric sweetener; a steviol glycoside that has been isolated from S. rebaudiana leaves; metabolized to steviol by gut microbiota