Learn More
TARGETMOL CHEMICALS INC REBAUDIOSIDE D 25MG
SDP
Catalog No. 502580437
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rebaudioside D is a potential sweetener. Rebaudioside D shows similar stability when exposed to simulate stomach and small intestine fluids with susceptibility to hydrolytic degradation by enteric bacteria collected from the cecum. purity: 99%
Catalog No. 50-258-0437 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S184025MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rebaudioside D is a potential sweetener. Rebaudioside D shows similar stability when exposed to simulate stomach and small intestine fluids with susceptibility to hydrolytic degradation by enteric bacteria collected from the cecum. purity: 99%