Learn More
Medchemexpress LLC Rebaudioside D | 63279-13-0 | 1129.15 | 1 ML
SDP
Catalog No. 5000389901
Encompass_Preferred
Rebaudioside D is an orally effective sweetener used in diet-induced obesity and related research. It activates Farnesoid X Receptor (FXR) and modulates lipid metabolism, reducing fatty acid and cholesterol synthesis.
  • Orally effective sweetener.
  • Activates Farnesoid X Receptor (FXR).
  • Modulates Acetyl-CoA Carboxylase and inhibits 3-hydroxy-3-methylglutaryl-CoA reductase.
  • Regulates bile acid homeostasis and lipid metabolism.
  • Reduces fatty acid and cholesterol synthesis.
  • Exhibits anti-adipogenesis, hepatoprotection, and anti-steatosis.
  • Modulates gut microbiota and enhances secondary bile acid metabolism.
  • Reduces body weight gain and visceral fat accumulation.
  • Used in diet-induced obesity and related research.
  • Does not affect glucose homeostasis or induce weight gain.
Catalog No. 50-003-89901 Supplier Medchemexpress LLC Supplier No. HYN046810MM/1ML
Rebaudioside D is an orally effective sweetener used in diet-induced obesity and related research. It activates Farnesoid X Receptor (FXR) and modulates lipid metabolism, reducing fatty acid and cholesterol synthesis.
  • Orally effective sweetener.
  • Activates Farnesoid X Receptor (FXR).
  • Modulates Acetyl-CoA Carboxylase and inhibits 3-hydroxy-3-methylglutaryl-CoA reductase.
  • Regulates bile acid homeostasis and lipid metabolism.
  • Reduces fatty acid and cholesterol synthesis.
  • Exhibits anti-adipogenesis, hepatoprotection, and anti-steatosis.
  • Modulates gut microbiota and enhances secondary bile acid metabolism.
  • Reduces body weight gain and visceral fat accumulation.
  • Used in diet-induced obesity and related research.
  • Does not affect glucose homeostasis or induce weight gain.