Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rebaudioside G | 127345-21-5 | 1 MG
SDP

Catalog No. 5000421029
Encompass_Preferred

Rebaudioside G is a minor constituent isolated from the leaves of *Stevia rebaudiana* Bertoni, used for sweeteners research. It is a solid with a colorless to off-white appearance.

  • Molecular Weight: 804.87
  • Chemical Formula: C38H60O18
  • Classification: Terpenoids and Diterpenoids
  • Initial Source: Plants other families
  • Purity: 99.89%
  • Solubility (In Vitro): H2O : 10 mg/mL (12.42 mM; Need ultrasonic and warming)

Catalog No. 50-004-21029 Supplier Medchemexpress LLC Supplier No. HYN22911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rebaudioside G is a minor constituent isolated from the leaves of *Stevia rebaudiana* Bertoni, used for sweeteners research. It is a solid with a colorless to off-white appearance.

  • Molecular Weight: 804.87
  • Chemical Formula: C38H60O18
  • Classification: Terpenoids and Diterpenoids
  • Initial Source: Plants other families
  • Purity: 99.89%
  • Solubility (In Vitro): H2O : 10 mg/mL (12.42 mM; Need ultrasonic and warming)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.