Learn More
Medchemexpress LLC Rebaudioside I | 1220616-34-1 | 5 MG
SDP
Catalog No. 5000417346
Encompass_Preferred
Rebaudioside I is a natural, non-caloric sweetener that is isolated from the *S. rebaudiana Morita* plant. This product is strictly for research use and is not intended for patient use or human consumption.
  • Natural origin: sourced from *S. rebaudiana Morita*.
  • Non-caloric: offers sweetness without caloric content.
  • High purity: guaranteed at 98.33% purity.
  • Research use only: specifically designed for laboratory and research applications.
  • Multiple pack sizes: available in various quantities including 1 mg, 5 mg, 10 mg, and 50 mg.
  • Comprehensive documentation: includes a data sheet, safety data sheet (SDS), certificate of analysis (COA), HNMR, and RP-HPLC.
Catalog No. 50-004-17346 Supplier Medchemexpress LLC Supplier No. HYN68895MG
Rebaudioside I is a natural, non-caloric sweetener that is isolated from the *S. rebaudiana Morita* plant. This product is strictly for research use and is not intended for patient use or human consumption.
  • Natural origin: sourced from *S. rebaudiana Morita*.
  • Non-caloric: offers sweetness without caloric content.
  • High purity: guaranteed at 98.33% purity.
  • Research use only: specifically designed for laboratory and research applications.
  • Multiple pack sizes: available in various quantities including 1 mg, 5 mg, 10 mg, and 50 mg.
  • Comprehensive documentation: includes a data sheet, safety data sheet (SDS), certificate of analysis (COA), HNMR, and RP-HPLC.