Learn More
Cayman Chemical Rebaudioside M, Purity: ≥95%, CAS: 1220616-44-3, 10 mg
SDP
Catalog No. 502909553
Encompass_Preferred
A natural non-caloric sweetener and TAS1R2/TAS1R3 agonist; increases intracellular calcium levels in HEK293 cells expressing human TAS1R2 and TAS1R3 (EC50 = 29.54 µM); has a relative sweetness potency 200-350 times that of sucrose; metabolized by gut microbiota to steviol
Catalog No. 50-290-9553 Supplier Cayman Chemical Supplier No. 3523910 MG
A natural non-caloric sweetener and TAS1R2/TAS1R3 agonist; increases intracellular calcium levels in HEK293 cells expressing human TAS1R2 and TAS1R3 (EC50 = 29.54 µM); has a relative sweetness potency 200-350 times that of sucrose; metabolized by gut microbiota to steviol